Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C-Reactive Protein/CRP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$616.37
Specifications
| Antigen | C-Reactive Protein/CRP |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20-1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
C-Reactive Protein/CRP Polyclonal specifically detects C-Reactive Protein/CRP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| C-Reactive Protein/CRP | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895 | |
| CRP | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20-1:50 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cytokine Research, Immunology, Innate Immunity | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 1401 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MSRKAFVFPKESDTSYVSLKAPLTKPLKAFTVCLHFYTELSSTRGYSIFSYATKRQDNEILIFWSKDIGYSFTVGGS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title