Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C-Reactive Protein/CRP Rabbit anti-Mouse, Clone: 32, Novus Biologicals™

Rabbit Monoclonal Antibody
$344.00 - $527.00
Specifications
| Antigen | C-Reactive Protein/CRP |
|---|---|
| Clone | 32 |
| Dilution | Sandwich ELISA Capture 1:250-1:2000 |
| Applications | ELISA |
| Classification | Monoclonal |
Description
C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Mouse samples. It is validated for Sandwich ELISA Capture.Specifications
| C-Reactive Protein/CRP | |
| Sandwich ELISA Capture 1:250-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895 | |
| Recombinant Mouse C-Reactive Protein/CRP Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 32 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 0.2 um filtered solution in PBS | |
| 1401 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title