Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ C-Reactive Protein, Mouse, Unlabeled, Clone: 2A8.1,

Catalog No. MABF1070MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF1070MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1070MI Supplier MilliporeSigma™ Supplier No. MABF1070

Mouse Monoclonal Antibody

C-reactive protein (UniProt P02741) is encoded by the CRP (also known as PTX1) gene (Gene ID 1401) in human. CRP is the classical acute phase reactant produced by the liver and its circulating concentration increases rapidly in a cytokine-mediated response to tissue injury, infection, and inflammation. It is an annular, pentameric protein that binds to lysophosphatidylcholine expressed on the surface of dead or dying cells and bacteria in order to activate the complement system via the C1Q complex. Interleukin-6 (IL-6), produced by macrophages in response to injury and infection, is a major trigger for the synthesis of CRP by the liver. It plays a major role in early host defense by promoting agglutination, bacterial capsular swelling, phagocytosis, and complement fixation through its calcium-dependent binding to phosphorylcholine. It can also interact with DNA and histones and scavenge nuclear material released from damaged circulating cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C-Reactive Protein
Applications Western Blot
Classification Monoclonal
Clone 2A8.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG2bκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene CRP, PTX1
Gene Accession No. P02741
Host Species Mouse
Immunogen GST-tagged recombinant human C-reactive protein C-terminal fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_000558
Test Specificity Clone 2A8.1 targets an epitope within the C-terminal half of human C-reactive protein.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.