Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
c-Rel Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10097834C
Description
c-Rel Polyclonal antibody specifically detects c-Rel in Human samples. It is validated for Western BlotSpecifications
c-Rel | |
Polyclonal | |
Western Blot | |
C-Rel, C-Rel proto-oncogene protein, I-Rel, oncogene REL, avian reticuloendotheliosis, proto-oncogene c-Rel, v-rel avian reticuloendotheliosis viral oncogene homolog, v-rel reticuloendotheliosis viral oncogene homolog (avian) | |
The immunogen recognized by this antibody maps to a region between residue 569 and 619 of human reticuloendotheliosis viral oncogene homolog using the numbering given in entry NP_002899.1 (GeneID 5966). | |
0.1 mL | |
Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors | |
5966 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction