Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C. rhodostoma Snaclec rhodocytin subunit alpha Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117723
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117723 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117723 Supplier Invitrogen™ Supplier No. PA5117723
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Elicits platelet aggregation by the binding to the C-type lectin domain family 1 member B (CLEC1B/CLEC2). Binding leads to tyrosine phosphorylation in the cytoplasmic tail of CLEC1B, which promotes the binding of spleen tyrosine kinase (Syk), subsequent activation of PLC-gamma-2, and platelet activation and aggregation. Binding to GPIbalpha (GP1BA) and alpha-2/beta-1 (ITGA2/ITGB1) may also induce aggregation, but this is controversial.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C. rhodostoma Snaclec rhodocytin subunit alpha
Applications Western Blot
Classification Polyclonal
Concentration 3.609 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Alias Rhodoaggretin subunit alpha
Host Species Rabbit
Immunogen Recombinant Calloselasma rhodostoma Snaclec rhodocytin subunit alpha protein (1-136aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Reptile
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.