Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
c-SRC Mouse anti-Hamster, Human, Mouse, Clone: 5E10C4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | c-SRC |
---|---|
Clone | 5E10C4 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6405
![]() ![]() |
Proteintech Group Inc
603151IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
SRC is a non-receptor kinase involved in the regulation of growth and differentiation of eukaryotic cells. SRC associates with the cytoplasmic region of various cell surface receptors to facilitate signal transduction, and is also involved in cytoskeletal organization, cell cycle regulation, and apoptosis. In humans, the gene encoding SRC is present on chromosome 20. Activity of SRC is regulated by two tyrosine residues (Tyr 416 and Tyr527), and phosphorylation of these two sites have opposing effects. Phosphorylation of Tyr416 in the activation loop of the kinase domain upregulates enzyme activity while the phosphorylation of Tyr527 decreases enzyme activity. Mutations in the SRC gene could be involved in the maligt progression of colon cancer. Two transcript variants encoding the same SRC protein have been found thus far.Specifications
c-SRC | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ASV, c SRC, c-SRC, p60 Src, pp60c src, Proto oncogene c Src, SRC, SRC1 | |
SRC | |
IgG2b | |
Protein A | |
Antibody |
5E10C4 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Hamster, Human, Mouse | |
P05480, P12931 | |
100757188, 20779, 6714 | |
c-SRC Fusion Protein Ag22306 | |
Primary | |
-20°C | |
SRC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title