Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C. tetani TeNT Light Chain Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6535SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6535SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6535SP Supplier R&D Systems Supplier No. MAB6535SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TeNT Light Chain/Tetanus Toxin Monoclonal specifically detects TeNT Light Chain/Tetanus Toxin in C. tetani samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen TeNT Light Chain/Tetanus Toxin
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 604023
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P04958
Gene Alias TeNT-LC, Tentoxylysin, Tetanus toxin chain H, Tetanus toxin chain L, Tetanus toxin heavy chain, Tetanus toxin light chain, tetX
Gene Symbols TETX
Host Species Mouse
Immunogen E. coli-derived recombinant C. tetani TeNT Light Chain Pro2-Gly430 Accession # P04958
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1061100
Test Specificity Detects TeNT Light Chain in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant TeNT Heavy Chain is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species C. tetani
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.