Learn More
Invitrogen™ C. tetani TeNT Light Chain Monoclonal Antibody (604023)

Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant TeNT Heavy Chain is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
Specifications
| Antigen | C. tetani TeNT Light Chain |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 604023 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene Alias | TeNT-LC; Tentoxylysin; tetX |
| Host Species | Mouse |
| Immunogen | E. coli-derived recombinant C. tetani TeNT Light Chain Pro2-Gly430. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.