Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1 inhibitor Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531541
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531541 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531541 Supplier Invitrogen™ Supplier No. PA531541
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: human plasma, mouse plasma, rat plasma. Predicted reactivity: Human (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

C1-inhibitor (C1INH) functions as a regulator of the activation of the classical complement pathway and of the contact activation system of kinin generation and coagulation. Its primary biologically relevant target proteinases are C1r, C1s, coagulation factors XIa and XIIa, and plasma kallikrein. C1-inhibitor consists of one polypeptide chain and has a molecular mass of ~104 kDa. Serum concentration has been determined to 180 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1 inhibitor
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene SERPING1
Gene Accession No. P05155, P97290, Q6P734
Gene Alias C1 esterase inhibitor; C1 Inh; C1 inhibitor; C1IN; C1Inh; C1-inhibiting factor; C1nh; complement component 1 inhibitor; complement component 1 inhibitor (angioedema, hereditary); esterase inhibitor; factor XIIa inhibitor; HAE1; HAE2; IC1; Plasma protease C1 inhibitor; serine (or cysteine) peptidase inhibitor, clade G, member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serine (or cysteine) proteinase inhibitor, clade G, member 1; serine/cysteine proteinase inhibitor clade G member 1; serpin family G member 1; serpin G1; serpin peptidase inhibitor clade G member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serpin peptidase inhibitor, clade G, member 1; SERPING1; XIIaINH
Gene Symbols SERPING1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human C1 inhibitor. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12258, 295703, 710
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.