Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1 inhibitor Recombinant Rabbit Monoclonal Antibody (0D3W8)
GREENER_CHOICE

Catalog No. PIMA537889
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537889 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537889 Supplier Invitrogen™ Supplier No. MA537889
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: Human plasma, Mouse plasma, Rat plasma Immunogen sequence: SLKLYHAFSA MKKVETNMAF SPFSIASLLT QVLLGAGENT KTNLESILSY PKDFTCVHQA LKGFTTKGVT SVSQIFHSPD LAIRDTFVNA SRTLYSSSPR V.

C1-inhibitor (C1INH) functions as a regulator of the activation of the classical complement pathway and of the contact activation system of kinin generation and coagulation. Its primary biologically relevant target proteinases are C1r, C1s, coagulation factors XIa and XIIa, and plasma kallikrein. C1-inhibitor consists of one polypeptide chain and has a molecular mass of ~104 kDa. Serum concentration has been determined to 180 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1 inhibitor
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 0D3W8
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
Gene SERPING1
Gene Accession No. P05155, P97290, Q6P734
Gene Alias C1 esterase inhibitor; C1 Inh; C1 inhibitor; C1IN; C1Inh; C1-inhibiting factor; C1nh; complement component 1 inhibitor; complement component 1 inhibitor (angioedema, hereditary); esterase inhibitor; factor XIIa inhibitor; HAE1; HAE2; IC1; Plasma protease C1 inhibitor; serine (or cysteine) peptidase inhibitor, clade G, member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serine (or cysteine) proteinase inhibitor, clade G, member 1; serine/cysteine proteinase inhibitor clade G member 1; serpin family G member 1; serpin G1; serpin peptidase inhibitor clade G member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serpin peptidase inhibitor, clade G, member 1; SERPING1; XIIaINH
Gene Symbols SERPING1
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12258, 295703, 710
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.