Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CABCOCO1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559550
Description
Immunogen sequence: EMKRLDQEQG PEESQPETDT SDMDPLVGFT IEDVKSVLDQ VTDDILIGIQ TEINEKLQIQ EEAFNARIE Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 67%.
CABCOCO1 (C10orf107) is a calcium-binding protein that may be involved in the control of sperm flagellar movement.
Specifications
CABCOCO1 | |
Polyclonal | |
Unconjugated | |
CABCOCO1 | |
bA63A2.1; C10orf107; CABCOCO1; chromosome 10 open reading frame 107; Ciliary-associated calcium-binding coiled-coil protein 1; RGD1306739; similar to RIKEN cDNA 1700040L02; uncharacterized protein C10orf107; uncharacterized protein C10orf107 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
219621 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IVU9 | |
CABCOCO1 | |
Recombinant protein corresponding to Human C10orf107. Recombinant protein control fragment (Product #RP-97194). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction