Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C10orf118 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553195
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553195 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553195 Supplier Invitrogen™ Supplier No. PA553195
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRSSTFPESA NEKTYSESPY DTDCTKKFIS KIKSVSASED LLEEIESELL STEFAEHRVP NGMNKGEHAL VLFEKCVQDK YLQQEHIIKK LIKENKKHQE LFVDICSEKD NLREELKKRT ETEK Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 73%.

CCDC186 (Coiled-Coil Domain Containing 186) is a Protein Coding gene. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C10orf118
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC186
Gene Accession No. Q7Z3E2
Gene Alias 1810028B20Rik; A630007B06Rik; AI159700; C10orf118; CCDC186; coiled-coil domain containing 186; coiled-coil domain-containing protein 186; CTCL tumor antigen HD-CL-01/L14-2; CTCL tumor antigen L14-2; FLJ10188; FLJ35301; oocyte-testis gene 1 protein; Otg1; RGD1307158
Gene Symbols CCDC186
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C10orf118. Recombinant protein control fragment (Product #RP-90821).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55088
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.