Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C10orf128 Polyclonal Antibody
C10orf128 Polyclonal Antibody
Supplier: Thermo Scientific PA563531
Description
C10orf128 Polyclonal Antibody for IHC

Specifications
C10orf128 | |
Polyclonal | |
Unconjugated | |
C10orf128 | |
C10orf128; chromosome 10 open reading frame 128; putative uncharacterized protein C10orf128; TMEM273; Transmembrane protein 273 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170371 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T292 | |
C10orf128 | |
Recombinant protein corresponding to Human C10orf128. Recombinant protein control fragment (Product #RP-104119). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction