Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C10orf132 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570010
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 92%; Rat: 100%; Zebrafish: 91%.
GOLGA7B gene ontology annotations related to this gene include peptidyl-L-cysteine S-palmitoylation; protein targeting to membrane.
Specifications
C10orf132 | |
Polyclonal | |
Unconjugated | |
GOLGA7B | |
4933417O08Rik; AI839934; bA451M19.3; bA459F3.4; C10orf132; C10orf133; GOLGA7B; golgi autoantigen, golgin subfamily a, 7B; golgin A7 family member B; golgin A7 family, member B; Golgin subfamily A member 7B | |
Rabbit | |
Affinity chromatography | |
RUO | |
401647 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q2TAP0 | |
GOLGA7B | |
synthetic peptide directed towards the middle region of human C10orf132 (aa 3-52). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction