Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C10orf32 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558040
Description
Immunogen sequence: FGQSVKGLLT EKVTTCGTDV IALTKQVLKG SRSSELLGQA ARNMVLQEDA ILHSEDSLRK MAIITTHLQY QQEAIQKNVE QSSDLQDQLN HLLK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
C10orf32 (chromosome 10 open reading frame 32) is a 105 amino acid protein that belongs to the UPF0693 family. The gene encoding C10orf32 maps to human chromosome 10, which spans nearly 135 million base pairs, makes up approximately 4.5% of total DNA in cells and encodes nearly 1,200 genes. Several protein-coding genes, including those that encode for chemokines, cadherins, excision repair proteins, early growth response factors (Egrs) and fibroblast growth receptors (FGFRs), are located on chromosome 10. Defects in some of the genes that map to chromosome 10 are associated with Charcot-Marie-Tooth disease, Jackson-Weiss syndrome, Usher syndrome, nonsyndromatic deafness, Wolman's syndrome, Cowden syndrome, multiple endocrine neoplasia type 2 and porphyria.
Specifications
C10orf32 | |
Polyclonal | |
Unconjugated | |
BORCS7 | |
2010012O05Rik; 4930569L17Rik; AI413851; AI467257; BLOC-1 related complex subunit 7; BLOC-1-related complex subunit 7; BORCS7; C10orf32; diaskedin; RGD1311783; RIKEN cDNA 2010012O05 gene; similar to RIKEN cDNA 2010012O05; UPF0693 protein C10orf32; UPF0693 protein C10orf32 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
119032 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96B45 | |
BORCS7 | |
Recombinant protein corresponding to Human C10orf32. Recombinant protein control fragment (Product #RP-97173). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction