Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C10orf4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530979
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530979 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530979 Supplier Invitrogen™ Supplier No. PA530979
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, HeLa nucleus. Predicted reactivity: Human (99%), Mouse (90%), Rat (91%), Pig (93%), Rhesus Monkey (98%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a nuclear phosphoprotein of unknown function. The 5'; UTR of this gene is part of a CpG island and contains a tandem CGG repeat region that normally consists of 8-14 repeats but can expand to over 200 repeats. The expanded allele becomes hypermethylated and is not transcribed; however, an expanded repeat region has not been associated with any disease phenotype. This gene is found within the rare FRA10A folate-sensitive fragile site.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C10orf4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene FRA10AC1
Gene Accession No. Q70Z53
Gene Alias 5730455O13Rik; AI448293; C10orf4; F26C11.1-like; FRA10A; Fra10ac1; FRA10AC1 homolog (human); fragile site 10q23.3; fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate 1; PRO2972; Protein FRA10AC1; protein FRA10AC1 homolog; rare folic acid-type fragile site, FRA(10)(q23.3), candidate gene 1; RGD1309482
Gene Symbols FRA10AC1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human C10orf4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118924
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.