Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C10orf46 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113995
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113995 Supplier Invitrogen™ Supplier No. PA5113995
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SELSEYAAQD QKFQRELIQN GFTRGDQSRK RAGDELAYNS SSACASSRGY For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Cell cycle associated protein capable of promoting cell proliferation through the activation of CDK2 at the G1/S phase transition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C10orf46
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CACUL1
Gene Accession No. Q86Y37
Gene Alias 2700078E11Rik; 2810417M16Rik; 9830127L17Rik; AI450346; C10orf46; CAC1; CACUL1; CDK2 associated cullin domain 1; CDK2 associated, cullin domain 1; CDK2-associated and cullin domain-containing protein 1; CDK2-associated, cullin domain 1; Cdk-associated cullin1; cullin; D130033C15Rik; RGD1308127
Gene Symbols CACUL1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human C10orf46.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143384
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.