Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C10orf46 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | C10orf46 |
---|---|
Concentration | 0.46 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5306
![]() ![]() |
Proteintech Group Inc
263211AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Cell cycle associated protein capable of promoting cell proliferation through the activation of CDK2 at the G1/S phase transition.Specifications
C10orf46 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q86Y37, Q8R0X2 | |
143384, 78832 | |
C10orf46 Fusion Protein Ag23694 | |
Primary | |
-20°C | |
CACUL1 |
0.46 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C10orf46, CACUL1 | |
Cacul1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title