Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C10orf47 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544380
Description
Peptide sequence: LCFRPGPALP STRARQSFPG PRQPNGAQDW RRADSLPRPQ GITVQFAGRG Sequence homology: Human: 100%.
The function of this protein remains unknown.
Specifications
C10orf47 | |
Polyclonal | |
Unconjugated | |
PROSER2 | |
5430407P10Rik; AU067770; C10orf47; proline and serine rich 2; proline and serine-rich protein 2; Proser2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
254427 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86WR7 | |
PROSER2 | |
Synthetic peptide directed towards the C-terminal of human C10orf47 (aa 366-415). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction