Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C10orf62 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304546100UL
Description
C10orf62 Polyclonal specifically detects C10orf62 in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
C10orf62 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
414157 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry | |
Unconjugated | |
bA548K23.1, chromosome 10 open reading frame 62 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-223 of human C10orf62 (NP_001009997.2). | |
100 μL | |
Primary | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction