Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C10orf76 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562264
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562264 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562264 Supplier Invitrogen™ Supplier No. PA562264
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEGKLESLDG EELMKIKDNI NCLFQHCIQA LGEEHPIRVV NALQTLCALI RGVHQKNKST SGFDIINMLM GFDKAELCMK NLM Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen C10orf76
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C10orf76
Gene Accession No. Q5T2E6
Gene Alias Armadillo-like helical domain-containing protein 3; ARMH3; C10orf76; chromosome 10 open reading frame 76; UPF0668 protein C10orf76
Gene Symbols C10orf76
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C10orf76. Recombinant protein control fragment (Product #RP-101233).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79591
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.