Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C10orf82 Antibody (OTI1G4), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272401AF350
Description
C10orf82 Monoclonal specifically detects C10orf82 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
C10orf82 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
C10orf82 | |
Full length recombinant protein of human C10orf82 (NP_653262) produced in HEK293T cells. | |
0.1 mL | |
143379.0 | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1G4 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
chromosome 10 open reading frame 82, Em:AC016825.4, FLJ40268, hypothetical protein LOC143379, MGC33547 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction