Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C10orf82 Antibody (OTI1G4) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272401
Description
C10orf82 Monoclonal specifically detects C10orf82 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
C10orf82 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
chromosome 10 open reading frame 82, Em:AC016825.4, FLJ40268, hypothetical protein LOC143379, MGC33547 | |
Mouse | |
17.6 kDa | |
100 μg | |
143379 | |
Human | |
Lyophilized |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1G4 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
C10orf82 | |
Full length human recombinant protein of human C10orf82 (NP_653262) produced in HEK293T cell. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction