Learn More
C10orf82 Mouse anti-Human, Clone: OTI1G4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501481

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
C10orf82 (Chromosome 10 Open Reading Frame 82) is a Protein Coding gene.Specifications
C10orf82 | |
Monoclonal | |
Unconjugated | |
C10orf82 | |
chromosome 10 open reading frame 82 | |
Mouse | |
Affinity Chromatography | |
RUO | |
143379 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1G4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8WW14 | |
C10orf82 | |
Full length human recombit protein of human C10orf82 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.