Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C10orf88 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558879
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558879 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558879 Supplier Invitrogen™ Supplier No. PA558879
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CSQVNHLHVG NKTECQENIT KHGERILGVG MEEQSICSYL EKILSKNMEL MEKKLMDYID QRIHELQEHI DDKIALLLDL LQNPNSP Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 68%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen C10orf88
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C10orf88
Gene Accession No. Q9H8K7
Gene Alias C10orf88; chromosome 10 open reading frame 88; uncharacterized protein C10orf88
Gene Symbols C10orf88
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C10orf88. Recombinant protein control fragment (Product #RP-97075).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80007
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.