Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C10orf90 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558480
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558480 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558480 Supplier Invitrogen™ Supplier No. PA558480
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEDTLFQAPP ALANGAHPGR HQRSFACTEF SRNSSVVRLK VPEAHTGLCE RRKYWVTHAD DKETSFSPDT PLSGKSPLVF SSCVHLRVSQ QCPD Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 62%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen C10orf90
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C10orf90
Gene Accession No. Q96M02
Gene Alias (E2-independent) E3 ubiquitin-conjugating enzyme FATS; bA422P15.2; C10orf90; Centrosomal protein C10orf90; chromosome 10 open reading frame 90; E2/E3 hybrid ubiquitin-protein ligase FATS; FATS; Fragile-site associated tumor suppressor homolog
Gene Symbols C10orf90
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C10orf90. Recombinant protein control fragment (Product #RP-97127).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118611
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.