Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C10orf96 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570165
Description
This target displays homology in the following species: Cow: 79%; Dog: 86%; Guinea Pig: 86%; Horse: 85%; Human: 100%; Pig: 93%; Rabbit: 86%; Yeast: 85%.
CCDC172 is a protein coding gene.
Specifications
C10orf96 | |
Polyclonal | |
Unconjugated | |
CCDC172 | |
C10orf96; C26H10orf96; C28H10orf96; CCDC172; coiled-coil domain containing 172; coiled-coil domain-containing protein 172; TEKT2-binding protein 1; Tekt2bp1; Unknown (protein for MGC:134210); UPF0628 protein C10orf96; UPF0628 protein C10orf96 homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
374355 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P0C7W6 | |
CCDC172 | |
synthetic peptide directed towards the middle region of human C10orf96. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction