Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C10orf97 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558376
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558376 Supplier Invitrogen™ Supplier No. PA558376
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSELTKELME LVWGTKSSPG LSDTIFCRWT QGFVFSESEG SALEQFEGGP CAVIAPVQAF LLKKLLFSSE KSSWRDCSEE EQKELLCHTL CDILESACCD HSGS Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

The protein encoded by this gene contains a caspase-associated recruitment domain and may function in apoptosis. It has been identified as a tumor suppressor in lung and gastric cancers, and a polymorphism in the gene may be associated with gastric cancer risk. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C10orf97
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MINDY3
Gene Accession No. Q9CV28, Q9H8M7
Gene Alias 1810041E18Rik; 2310047O13Rik; 5830410F13Rik; AI447827; AW111958; C10orf97; CARD-containing protein; Carp; caspase recruitment domain containing pro-apoptotic protein; caspase recruitment domain containing protein; dermal papilla-derived protein 5; DERP5; Deubiquitinating enzyme MINDY-3; Fam188a; family with sequence similarity 188 member A; family with sequence similarity 188, member A; MINDY3; MST126; MSTP126; My042; Protein CARP; protein FAM188A; RGD1309605; RP11-394I23.1; Ubiquitin carboxyl-terminal hydrolase MINDY-3
Gene Symbols Fam188a, MINDY3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C10orf97. Recombinant protein control fragment (Product #RP-97100).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291320, 66960, 80013
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.