Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C10orf97 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570591
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570591 Supplier Invitrogen™ Supplier No. PA570591
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 100%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%.

The protein encoded by this gene contains a caspase-associated recruitment domain and may function in apoptosis. It has been identified as a tumor suppressor in lung and gastric cancers, and a polymorphism in the gene may be associated with gastric cancer risk. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C10orf97
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Fam188a
Gene Accession No. 0
Gene Alias 1810041E18Rik; 2310047O13Rik; 5830410F13Rik; AI447827; AW111958; C10orf97; CARD-containing protein; Carp; caspase recruitment domain containing pro-apoptotic protein; caspase recruitment domain containing protein; dermal papilla-derived protein 5; DERP5; Deubiquitinating enzyme MINDY-3; Fam188a; family with sequence similarity 188 member A; family with sequence similarity 188, member A; MINDY3; MST126; MSTP126; My042; Protein CARP; protein FAM188A; RGD1309605; RP11-394I23.1; Ubiquitin carboxyl-terminal hydrolase MINDY-3
Gene Symbols Fam188a
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of rat C10orf97.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291320
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.