Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C11orf1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP312205
This item is not returnable.
View return policy
Description
C11orf1 Polyclonal antibody specifically detects C11orf1 in Human samples. It is validated for ELISASpecifications
C11orf1 | |
Polyclonal | |
Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide | |
Rabbit | |
Affinity-purified | |
RUO | |
64776 | |
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
ELISA | |
Unconjugated | |
chromosome 11 open reading frame 1, FLJ23499, hypothetical protein LOC64776 | |
Mid-region epitope synthetic peptide taken within aino acid region 50-100 on human C11orf1 protein | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction