Learn More
Invitrogen™ C11orf24 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139743
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: VATTANTSSP MSTRPSPSKH MPSDTAASPV PPMRPQAQGP ISQVSVDQPV VNTTNKSTPM PSNTTPEPAP TPTVVTTTKA QAREPTASPV PVPHTSPIPE MEAMSPTTQP SPMPYTQRAA GPGTSQAPEQ VETEATPGT.
The function remains unknown.
Specifications
C11orf24 | |
Polyclonal | |
Unconjugated | |
C11orf24 | |
C11orf24; chromosome 11 open reading frame 24; DM4E3; FP2568; Protein DM4E3; Uncharacterized protein C11orf24; UNQ1872/PRO4315 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
53838 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96F05 | |
C11orf24 | |
Recombinant protein corresponding to Human C11orf24. Recombinant protein control fragment (Product #RP-90466). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.