Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C11orf24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543405
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543405 Supplier Invitrogen™ Supplier No. PA543405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SPVTLTKGTS AAHLNSMEVT TEDTSRTDVS EPATSGGAAD GVTSIAPTAV Sequence homology: Human: 100%.

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C11orf24
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C11orf24
Gene Accession No. Q96F05
Gene Alias C11orf24; chromosome 11 open reading frame 24; DM4E3; FP2568; Protein DM4E3; Uncharacterized protein C11orf24; UNQ1872/PRO4315
Gene Symbols C11orf24
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human C11orf24 (aa 61-110).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53838
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.