Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C11orf30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102867
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102867 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102867 Supplier Invitrogen™ Supplier No. PA5102867
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total C11orf30.

EMSY is a regulator which is able to repress transcription, possibly via its interaction with a multiprotein chromatin remodeling complex that modifies the chromatin. Its interaction with BRCA2 suggests that it may play a central role in the DNA repair function of BRCA2. As part of a histone H3-specific methyltransferase complex may mediate ligand-dependent transcriptional activation by nuclear hormone receptors. Diseases associated with EMSY include Sporadic Breast Cancer and Breast Cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C11orf30
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EMSY
Gene Accession No. Q7Z589
Gene Alias 2210018M11Rik; 2310016L09Rik; AU019521; AU019557; BRCA2-interacting transcriptional repressor EMSY; C11orf30; EMSY; EMSY BRCA2-interacting transcriptional repressor; EMSY protein; EMSY, BRCA2 interacting transcriptional repressor; EMSY, BRCA2-interacting transcriptional repressor; FLJ90741; GL002; protein EMSY; RGD1310712; RIKEN cDNA 2210018M11 gene; similar to EMSY protein
Gene Symbols EMSY
Host Species Rabbit
Immunogen A synthesized peptide derived from human EMSY(Accession Q7Z589), corresponding to amino acid residues Q414-Q464.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56946
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.