Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C11orf30 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576850
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
EMSY is a regulator which is able to repress transcription, possibly via its interaction with a multiprotein chromatin remodeling complex that modifies the chromatin. Its interaction with BRCA2 suggests that it may play a central role in the DNA repair function of BRCA2. As part of a histone H3-specific methyltransferase complex may mediate ligand-dependent transcriptional activation by nuclear hormone receptors. Diseases associated with EMSY include Sporadic Breast Cancer and Breast Cancer.
Specifications
C11orf30 | |
Polyclonal | |
Unconjugated | |
EMSY | |
2210018M11Rik; 2310016L09Rik; AU019521; AU019557; BRCA2-interacting transcriptional repressor EMSY; C11orf30; EMSY; EMSY BRCA2-interacting transcriptional repressor; EMSY protein; EMSY, BRCA2 interacting transcriptional repressor; EMSY, BRCA2-interacting transcriptional repressor; FLJ90741; GL002; protein EMSY; RGD1310712; RIKEN cDNA 2210018M11 gene; similar to EMSY protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
233545, 361607, 56946 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z589, Q8BMB0 | |
EMSY | |
Recombinant full length Human C11orf30. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction