Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C11orf42 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564116
Description
Immunogen sequence: QLLRLLRSLP VAFSCLKFSL QSKGVLGPQK PLTKDPLPHG ANWVRPNLSI MPPLAPTSAP AD Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 82%.
C11orf42 is a protein coding gene.
Specifications
C11orf42 | |
Polyclonal | |
Unconjugated | |
C11orf42 | |
C11orf42; chromosome 11 open reading frame 42; uncharacterized protein C11orf42 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
160298 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N5U0 | |
C11orf42 | |
Recombinant protein corresponding to Human C11orf42. Recombinant protein control fragment (Product #RP-103511). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction