Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C11orf42 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569932
Description
This target displays homology in the following species: Cow: 86%; Dog: 79%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 79%; Rabbit: 79%; Rat: 86%.
C11orf42 is a protein coding gene.
Specifications
C11orf42 | |
Polyclonal | |
Unconjugated | |
C11orf42 | |
C11orf42; chromosome 11 open reading frame 42; uncharacterized protein C11orf42 | |
Rabbit | |
Affinity chromatography | |
RUO | |
160298 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N5U0 | |
C11orf42 | |
synthetic peptide directed towards the middle region of human C11orf42 (aa 259-308). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction