Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C11orf46 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561661
Description
Immunogen sequence: FSGMDLCDCL DEDCLGCFYA CPACGSTKCG AECRCDRKWL YEQIEIEGGE IIHNKHA Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Specifications
C11orf46 | |
Polyclonal | |
Unconjugated | |
ARL14EP | |
2700007P21Rik; 4930448O08Rik; ADP ribosylation factor like GTPase 14 effector protein; ADP-ribosylation factor like GTPase 14 effector protein; ADP-ribosylation factor-like 14 effector protein; ARF7 effector protein; ARF7EP; ARL14 effector protein; ARL14EP; C11orf46; dJ299F11.1; RGD1311463 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
120534 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N8R7 | |
ARL14EP | |
Recombinant protein corresponding to Human C11orf46. Recombinant protein control fragment (Product #RP-97403). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction