Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C11ORF46 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989464
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9464 Supplier Bioss Supplier No. BS9932R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen C11ORF46
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ARL14EP
Gene Accession No. Q8N8R7
Gene Alias ADP ribosylation factor like GTPase 14 effector protein; ADP-ribosylation factor like GTPase 14 effector protein; ADP-ribosylation factor-like 14 effector protein; ARF7 effector protein; ARF7EP; ARL14 effector protein; ARL14EP; C11orf46; dJ299F11.1
Gene Symbols ARL14EP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C11ORF46.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120534
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.