Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C11orf49 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559132
Description
Immunogen sequence: VIVPTSSSGQ HRQRPALGGA GTLEGVEASL FYQCLENLCD RHKYSCPPPA LVKEALSNVQ RLTFYGFLMA LSKHRGINQA L Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.

Specifications
C11orf49 | |
Polyclonal | |
Unconjugated | |
C11orf49 | |
C11orf49; chromosome 11 open reading frame 49; RGD1309540; similar to hypothetical protein MGC40841; similar to hypothetical protein MGC4707; UPF0705 protein C11orf49; UPF0705 protein C11orf49 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79096 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H6J7 | |
C11orf49 | |
Recombinant protein corresponding to Human C11orf49. Recombinant protein control fragment (Product #RP-97370). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction