Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C11orf53 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | C11orf53 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15759420
![]() |
Novus Biologicals
NBP15759420UL |
20 μL |
Each for $206.00
|
|
|||||
NBP157594
![]() |
Novus Biologicals
NBP157594 |
100 μL |
Each for $487.50
|
|
|||||
Description
C11orf53 Polyclonal specifically detects C11orf53 in Human samples. It is validated for Western Blot.Specifications
C11orf53 | |
Polyclonal | |
Rabbit | |
Q8IXP5 | |
341032 | |
Synthetic peptides corresponding to C11ORF53 The peptide sequence was selected from the middle region of C11ORF53. Peptide sequence SIAQHRGSSWGSSLAGAQSYSLHALEDLHHTPGYPTPPPYPFTPFMTVSN. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
chromosome 11 open reading frame 53, hypothetical protein LOC341032, MGC50104 | |
C11ORF53 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title