Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C11orf53 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$331.00
Specifications
Antigen | C11orf53 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
C11orf53 Polyclonal specifically detects C11orf53 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
C11orf53 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
chromosome 11 open reading frame 53, hypothetical protein LOC341032, MGC50104 | |
C11ORF53 | |
IgG | |
Affinity Purified |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Human | |
Q8IXP5 | |
341032 | |
This antibody was developed against a recombinant protein corresponding to amino acids: STSCLSQLESGSIAQHRGSSWGSSLAGAQSYSLHALEDLHHTPGYPTPPPYPFTPFMTVSNDLPPKV | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title