Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C11orf53 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113095
Description
C11orf53 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The specific function of C11orf53 remains unknown.
Specifications
| C11orf53 | |
| Polyclonal | |
| Unconjugated | |
| C11orf53 | |
| C11orf53; chromosome 11 open reading frame 53; uncharacterized protein C11orf53 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 341032 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.58 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8IXP5 | |
| C11orf53 | |
| Recombinant Human Uncharacterized protein C11orf53 protein (13-236AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction