Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C11orf54 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542029
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542029 Supplier Invitrogen™ Supplier No. PA542029
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: CPDLTKEPFT FPVKGICGKT RIAEVGGVPY LLPLVNQKKV YDLNKIAKEI Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%; Zebrafish: 79%.

C11orf54, also known as Chromosome 11 Open Reading Frame 54, is an ester hydrolase highly conserved across various species. Located on chromosome 11, the gene is identified as a biomarker associated with renal cancers, although its precise biological function is not fully elucidated. Research has shown that C11orf54 plays a role in DNA repair mechanisms, particularly by blocking chaperone-mediated autophagy (CMA). Silencing this gene reduces cellular proliferation and heightens DNA damage and apoptosis when cells are treated with cisplatin, a common chemotherapeutic agent. This suggests C11orf54's involvement in cancer pathology, particularly its potential impact on DNA repair pathways, thereby supporting cell survival post DNA damage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C11orf54
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C11orf54
Gene Accession No. Q9H0W9
Gene Alias C11orf54; chromosome 11 open reading frame 54; ester hydrolase C11orf54; Ester hydrolase C11orf54 homolog; LP4947; PTD012; PTOD012; RGD1309534; similar to RIKEN cDNA 4931406C07
Gene Symbols C11orf54
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human C11orf54 (aa 38-87).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28970
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.