Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C11orf54 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559202
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559202 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559202 Supplier Invitrogen™ Supplier No. PA559202
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLGFNSEFMP VIQTESEHKP PVNGSYFAHV NPADGGCLLE KYSEKCHDFQ CALLANLFAS EGQPGKVIEV KAKRRTGPLN Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 80%.

C11orf54, also known as Chromosome 11 Open Reading Frame 54, is an ester hydrolase highly conserved across various species. Located on chromosome 11, the gene is identified as a biomarker associated with renal cancers, although its precise biological function is not fully elucidated. Research has shown that C11orf54 plays a role in DNA repair mechanisms, particularly by blocking chaperone-mediated autophagy (CMA). Silencing this gene reduces cellular proliferation and heightens DNA damage and apoptosis when cells are treated with cisplatin, a common chemotherapeutic agent. This suggests C11orf54's involvement in cancer pathology, particularly its potential impact on DNA repair pathways, thereby supporting cell survival post DNA damage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C11orf54
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C11orf54
Gene Accession No. Q9H0W9
Gene Alias C11orf54; chromosome 11 open reading frame 54; ester hydrolase C11orf54; Ester hydrolase C11orf54 homolog; LP4947; PTD012; PTOD012; RGD1309534; similar to RIKEN cDNA 4931406C07
Gene Symbols C11orf54
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C11orf54. Recombinant protein control fragment (Product #RP-97516).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28970
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.