Learn More
Invitrogen™ C11orf57 Polyclonal Antibody

Description
C11orf57 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | C11orf57 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | Nkapd1 |
| Gene Accession No. | Q6ZUT1 |
| Gene Alias | AU019823; C11orf57; chromosome 11 open reading frame 57; expressed sequence AU019823; Gm639; NKAP domain containing protein 1; NKAPD1; uncharacterized protein C11orf57; uncharacterized protein C11orf57 homolog; Uncharacterized protein NKAPD1 |
| Gene Symbols | Nkapd1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.