Learn More
Invitrogen™ C11orf57 Polyclonal Antibody

Description
Immunogen sequence: RWGHSGYKEL YPEEFETDSS DQQDITNGKK TSPQVKSSTH ESRKHKKSKK For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 100%.
Specifications
Specifications
| Antigen | C11orf57 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | Nkapd1 |
| Gene Accession No. | Q6ZUT1 |
| Gene Alias | AU019823; C11orf57; chromosome 11 open reading frame 57; expressed sequence AU019823; Gm639; NKAP domain containing protein 1; NKAPD1; uncharacterized protein C11orf57; uncharacterized protein C11orf57 homolog; Uncharacterized protein NKAPD1 |
| Gene Symbols | Nkapd1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.