Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C11orf57 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501733304 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3304 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3304 Supplier Proteintech Group Inc Supplier No. 211811AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C11orf57 has three isoforms with MW 30-34 kDa.

Specifications

Antigen C11orf57
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene C11orf57
Gene Accession No. Q6ZUT1
Gene Alias C11orf57
Gene Symbols C11orf57, Nkapd1
Host Species Rabbit
Immunogen C11orf57 Fusion Protein Ag15471
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 270156, 55216
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.