Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C11orf65 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569906
Description
This target displays homology in the following species: Horse: 77%; Human: 100%; Pig: 86%.
C11orf65 is a protein coding gene.
Specifications
C11orf65 | |
Polyclonal | |
Unconjugated | |
C11orf65 | |
C11orf65; chromosome 11 open reading frame 65; uncharacterized protein C11orf65 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
160140 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NCR3 | |
C11orf65 | |
synthetic peptide directed towards the N-terminal region of human C11orf65. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction