Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C11orf71 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567362
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567362 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567362 Supplier Invitrogen™ Supplier No. PA567362
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRPSVRAESRR VDGGGRSPRE PDGRGRSRQA RFSPYPIPAV EPDLLRSVL Highest antigen sequence identity to the following orthologs - mouse 68%, rat 66%.

C11orf71 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C11orf71
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C11orf71
Gene Accession No. Q6IPW1
Gene Alias C11orf71; chromosome 11 open reading frame 71; uncharacterized protein C11orf71; up-regulated in lung cancer 7; URLC7
Gene Symbols C11orf71
Host Species Rabbit
Immunogen Recombinant Human C11orf71. Recombinant protein control fragment (Product #RP-105667).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54494
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.