Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C11orf73 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100263
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100263 Supplier Invitrogen™ Supplier No. PA5100263
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Hikeshi.

Required for organization and/or function of the secretory apparatus in Clara cells in lung.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C11orf73
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HIKESHI
Gene Accession No. Q53FT3, Q5M808, Q9DD02
Gene Alias 0610007P06Rik; 1110002N09Rik; C11orf73; C1H11orf73; C21H11orf73; C29H11orf73; chromosome unknown open reading frame, human C11orf73; heat shock protein nuclear import factor hikeshi; Hikeshi; Hikeshi, heat shock protein nuclear import factor; HLD13; HSPC138; HSPC179; HSPC248; hypothetical protein LOC504867; hypothetical protein LOC550239; l(7)6Rn; l7Rn6; lethal gene on chromosome 7 Rinchik 6 protein; lethal, Chr 7, Rinchik 6; LOC100762248; OPI10; protein Hikeshi; Protein of unknown function DUF775 containing protein; similar to hypothetical protein HSPC138; uncharacterized protein C11orf73 homolog; Uncharacterized protein C11orf73-like; unknown; zgc:110091
Gene Symbols HIKESHI
Host Species Rabbit
Immunogen A synthesized peptide derived from human HIKESHI(Accession Q53FT3), corresponding to amino acid residues V66-E116.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293103, 51501, 67669
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.